Learn More
Strem, An Ascensus Company CAS# 12081-22-0. 2g. (1-Methylallyl)palladium chloride dimer, 99%. MFCD01074457
SDP
Supplier:  Strem, An Ascensus Company 4602752G
Encompass
CAS# 12081-22-0. 2g. (1-Methylallyl)palladium chloride dimer, 99%. MFCD01074457. Molecular Weight: 393.94. Molecular Formula: [C4H7PdCl]2. Color/form: yellow pwdr. Strem# 46-0275. http://www.strem.com/catalog/v/46-0275/
Catalog No. 50-901-12316