Learn More
Enterprise Technology Solutions Crown Mats & Matting Entrance/Antifatigue Mat, Rubber, 36"x72", Black
SDP
Catalog No. 5000790749
Encompass_Preferred
Rubber fingertip scraper mat features 60 percent more fingers than standard fingertip mats for increased scraping action. Durable design keeps its flexibility and maintains performance in cold temperatures. Beveled edge reservoir retains up to one gallon of liquid per square yard for an effective way to keep floors clean. Mat is designed for medium-traffic areas.
Catalog No. 50-007-90749 Supplier Enterprise Technology Solutions Supplier No. CWNMAFG62BK
Rubber fingertip scraper mat features 60 percent more fingers than standard fingertip mats for increased scraping action. Durable design keeps its flexibility and maintains performance in cold temperatures. Beveled edge reservoir retains up to one gallon of liquid per square yard for an effective way to keep floors clean. Mat is designed for medium-traffic areas.