Learn More
Chiral Technologies, Inc. CROWNPAK CR (-) HPLC ANALYTICA
Catalog No. NC9211710
Encompass
CROWNPAK CR, Crownpak CR(-), 5 micron, 4.0mm ID x 150mm Analytical column
Catalog No. NC9211710 Supplier Chiral Technologies, Inc. Supplier No. 28714
Add to Cart
Add to Cart
CROWNPAK CR, Crownpak CR(-), 5 micron, 4.0mm ID x 150mm Analytical column