Learn More
Chiral Technologies, Inc. CROWNPAKCR(+) 150X4.0MM
Supplier:  Chiral Technologies, Inc. 27714
Encompass
Crownpak CR Column, Particle Size: 5 micrometer, I.D.: 4mm, Column Length: 150mm, Product Type: Analytical, Absorbent: Crown ether.
Catalog No. NC9974510