Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Crozbaciclib fumarate | 2567494-39-5 | 99.4% | 604.65 | 10 MG
SDP

Catalog No. 5000435886
Encompass_Preferred

Crozbaciclib fumarate is a potent CDK4/6 inhibitor, demonstrating significant anti-proliferative effects by arresting the U87MG cell line exclusively in G1. It has shown promising results in vivo, including substantial tumor growth inhibition in orthotopic xenograft mouse models of glioblastoma multiforme.

  • Potent CDK4/6 inhibitor with IC50s of 3 nM (CDK4) and 1 nM (CDK6).
  • Acts as an anti-proliferative agent, arresting U87MG cells in G1.
  • Exhibits tumor growth inhibition from 62% to 99% in glioblastoma multiforme xenograft models.
  • Increases lifespan by 162% in mice at a dose of 50 mg/kg.

Catalog No. 50-004-35886 Supplier Medchemexpress LLC Supplier No. HY112280A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crozbaciclib fumarate is a potent CDK4/6 inhibitor, demonstrating significant anti-proliferative effects by arresting the U87MG cell line exclusively in G1. It has shown promising results in vivo, including substantial tumor growth inhibition in orthotopic xenograft mouse models of glioblastoma multiforme.

  • Potent CDK4/6 inhibitor with IC50s of 3 nM (CDK4) and 1 nM (CDK6).
  • Acts as an anti-proliferative agent, arresting U87MG cells in G1.
  • Exhibits tumor growth inhibition from 62% to 99% in glioblastoma multiforme xenograft models.
  • Increases lifespan by 162% in mice at a dose of 50 mg/kg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.