Learn More
Medchemexpress LLC Crozbaciclib fumarate | 2567494-39-5 | 99.4% | 604.65 g/mol | C32H34F2N6O4 | 5 MG
SDP
Catalog No. 5000264230
Encompass_Preferred
Crozbaciclib fumarate is the fumarate salt of crozbaciclib, a small-molecule cyclin-dependent kinase inhibitor selective for CDK4 and CDK6, provided for laboratory research. It exhibits low-nanomolar biochemical potency (CDK4 IC50 ≈ 3 nM; CDK6 IC50 ≈ 1 nM) and demonstrates antiproliferative activity in cellular assays.
  • Fumarate salt form for improved chemical stability.
  • Potent inhibition of CDK4 and CDK6 (low-nanomolar IC50s).
  • High chemical purity (reported 99.4%).
  • Supplied in small milligram quantities for research use.
Catalog No. 50-002-64230 Supplier Medchemexpress LLC Supplier No. HY112280A5MG
Crozbaciclib fumarate is the fumarate salt of crozbaciclib, a small-molecule cyclin-dependent kinase inhibitor selective for CDK4 and CDK6, provided for laboratory research. It exhibits low-nanomolar biochemical potency (CDK4 IC50 ≈ 3 nM; CDK6 IC50 ≈ 1 nM) and demonstrates antiproliferative activity in cellular assays.
  • Fumarate salt form for improved chemical stability.
  • Potent inhibition of CDK4 and CDK6 (low-nanomolar IC50s).
  • High chemical purity (reported 99.4%).
  • Supplied in small milligram quantities for research use.