Learn More
Enzo Life Sciences CRP (human) ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ENZKIT1020001
Encompass_Preferred
The CRP (human) ELISA kit is a complete, colorimetric, competitive immunoassay kit for the quantitative determination of human CRP in serum and plasma with results in 3 hours. Alternative name: C-reactive protein. Assay Time: 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-201-3806