Learn More
Invitrogen™ Human CRP Instant ELISA™ Kit
Description
The Human C-Reactive Protein (Hu CRP) ELISA quantitates Hu CRP in human serum, plasma and cell culture supernatants. The assay will exclusively recognize both natural and recombinant Hu CRP. Principle of the method The Human CRP solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P02741 |
| Assay Range | 78-5000 pg/mL |
| Assay Sensitivity | 3.0 pg/mL |
| Content And Storage | Aluminum pouch(es) with a Microplate coated with Polyclonal Antibody (murine) to human C-reactive protein, HRP-Conjugate (anti human C-reactive protein monoclonal (murine) antibody) and Assay Buffer, lyophilized Aluminum pouch(es) with a human C-reactive protein Standard curve (coloured) Wash Buffer Concentrate 20x (phosphate-buffered saline with 1% Tween 20) Substrate Solution (tetramethyl-benzidine) Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA) Stop Solution (1M Phosphoric acid) Adhesive Plate Covers |
| Conjugate | HRP |
| Immunoassay Kit Format | Coated ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.