Learn More
Invitrogen™ Rat CRP ELISA Kit
Description
The Rat C-Reactive Protein (Rt CRP) ELISA quantitates Rt CRP in rat serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt CRP. Principle of the method The Rat CRP solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P48199 |
| Assay Range | 0.246-60 ng/mL |
| Assay Sensitivity | 200 pg/mL |
| Calibration Range | 0.2 ng/mL to 60 ng/mL |
| Content And Storage | Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability. |
| Description | CRP (C-reactive) protein ELISA Kit, 96 Assays, 1 Kit, Rat Species, Proteases, Peptidases and Inhibitors Protein Family, Immunology Research Discipline, Enzyme-Linked Immunosorbent Assay Designed for the Quantitation and Detection of C-Reactive Protein (CR |
| Detectable Analytes | C-reactive protein (CRP) |
| Assay | Solid-phase sandwich ELISA |
| Conjugate | HRP |
| Detection Method | Colorimetric |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.