Learn More
Electron Microscopy Sciences Pipette Tip Crystal/Eppendorf, 0.1-10 mcL
SDP
Supplier:  Electron Microscopy Sciences 7244201
Encompass_Preferred
Pipette tips are low binding to ensure accuracy and precision. The low binding surface is the result of a manufacturing process that binds a perfluorooctyl polymer to polypropylene resin–resulting in an exposed trifluoromethyl surface. This surface reduces the surface energy of liquid and repels any substance so sample retention and binding are minimized.
Not detectable/Non-reactive.
Extracted only by a fluorinated solvent.
Autoclaving will not compromise performance.
Available in convenient hinged racked package.
Tips and racks are fully autoclavable.
Range: 0.1-10 µl
Sterile: Non-sterile
Catalog No. 50-189-9908