Learn More
Enzo Life Sciences Calreticulin monoclonal antibody (FMC 75) (PE conjugate) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS668PE0200
Encompass_Preferred
Clone: FMC 75. Isotype: IgG1κ. Immunogen: Recombinant calreticulin. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Monkey. Applications: Flow Cytometry. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide.
Catalog No. 50-201-3195