Learn More
Medchemexpress LLC HY-W014622 50mg Medchemexpress, CRT0044876 CAS:6960-45-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW01462250MG
Encompass_Preferred
Medchemexpress, HY-W014622 50mg CRT0044876 CAS:6960-45-8 CRT0044876 is a potent and selective apurinic/apyrimidinic endonuclease 1 (APE1) inhibitor (IC50=~3 μM). CRT0044876 inhibits the AP endonuclease, 3′-phosphodiesterase and 3′-phosphatase activities of APE1, and is a specific inhibitor of the exonuclease III family of enzymes to which APE1 belongs. CRT0044876 potentiates the cytotoxicity of several DNA base-targeting compounds. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3365