Learn More
Cayman Chemical CRT0066101hydrochlorIde 10mg
SDP
Catalog No. 502969806
Encompass_Preferred
A pan inhibitor PKD (IC50s = 1, 2.5, and 2 nM for PKD1, PKD2, and PKD3, respectively); blocks cell proliferation, induces apoptosis, and reduces the viability of pancreatic cancer cells both in vitro and in vivo
Catalog No. 50-296-9806 Supplier Cayman Chemical Supplier No. 1533710 MG
A pan inhibitor PKD (IC50s = 1, 2.5, and 2 nM for PKD1, PKD2, and PKD3, respectively); blocks cell proliferation, induces apoptosis, and reduces the viability of pancreatic cancer cells both in vitro and in vivo