Learn More
ACROBiosystems ACROBiosystems Human CRTAM Protein, His Tag
Catalog No. 501956724
Encompass_Preferred
Recombinant Protein;1MG;Human CRTAM, His Tag (CRM-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 18 - Gly 287 (Accession # O95727-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 31.9 kDa. The protein migrates as 42-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6724 Supplier ACROBiosystems Supplier No. CRMH52H31MG
Recombinant Protein;1MG;Human CRTAM, His Tag (CRM-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 18 - Gly 287 (Accession # O95727-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 31.9 kDa. The protein migrates as 42-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.