Learn More
ACROBiosystems Recombinant Protein;Cynomolgus CRTAM Protein, Fc Tag;HEK293;1MG;CRM,CRTAM,CRTA
Catalog No. 501659118
Encompass_Preferred
Recombinant Protein;Cynomolgus CRTAM Protein, Fc Tag;1MG;Cynomolgus CRTAM, Fc Tag (CRM-C5255) is expressed from human 293 cells (HEK293). It contains AA Ser 18 - Gly 287 (Accession # XP_005580021.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 56.6 kDa. The protein migrates as 85-95 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-9118 Supplier ACROBiosystems Supplier No. CRMC52551MG
Recombinant Protein;Cynomolgus CRTAM Protein, Fc Tag;1MG;Cynomolgus CRTAM, Fc Tag (CRM-C5255) is expressed from human 293 cells (HEK293). It contains AA Ser 18 - Gly 287 (Accession # XP_005580021.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 56.6 kDa. The protein migrates as 85-95 kDa under reducing (R) condition (SDS-PAGE).