Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci CRTC2 Peptide
SDP

Catalog No. 16597089
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-597-089 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-597-089 Supplier ProSci Supplier No. 7325P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

CRTC2 peptide is used for blocking the activity of CRTC2 antibody (catalog number 7325). It usually blocks the antibody activity completely in Western blot by incubating the peptide with equal volume of antibody for 30 min at 37°C.

Specifications

Gene ID (Entrez) 200186
Species Human
Accession Number NP_859066
Common Name CREB regulated transcription coactivator 2
Concentration 200 μg/mL
Content And Storage Store CRTC2 peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol CRTC2
Quantity 0.05 mg
Sequence 16 amino acid peptide near the carboxy terminus of human CRTC2.
Synonym TORC2, TORC-2, TORC2, CREB-regulated transcription coactivator 2, Transducer of regulated cAMP response element-binding protein 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.