Learn More
Cayman Chemical CRTH2 DP2 Receptor CTerm Pol
SDP
Catalog No. 502886547
Encompass_Preferred
Immunogen: Synthetic peptide from the C-terminal region of human CRTH2/DP2 • Host: Rabbit • Species Reactivity: (+) Human, mouse, and rat • Application(s): ICC and WB
Catalog No. 50-288-6547 Supplier Cayman Chemical Supplier No. 100070021 EA
Immunogen: Synthetic peptide from the C-terminal region of human CRTH2/DP2 • Host: Rabbit • Species Reactivity: (+) Human, mouse, and rat • Application(s): ICC and WB