Learn More
Cayman Chemical CRTH2/DP2 Receptor (N-Term) Blocking Peptide, 1 ea
SDP
Catalog No. 502887067
Encompass_Preferred
To be used in conjunction with Cayman’s CRTH2/DP2 Receptor (N-Term) polyclonal antibody (Item No. 10004886) to block protein-antibody complex formation during immunochemical analysis of CRTH2/DP2
Catalog No. 50-288-7067 Supplier Cayman Chemical Supplier No. 100048841 EA
To be used in conjunction with Cayman’s CRTH2/DP2 Receptor (N-Term) polyclonal antibody (Item No. 10004886) to block protein-antibody complex formation during immunochemical analysis of CRTH2/DP2