Learn More
Med Vet International Cruciate Needle, J0710M
Catalog No. 5000582202
Encompass_Preferred
20mm Small Cruciate Needle. Swaged onto 80 lb. nylon leader line with 12mm crimp tube. 800mm L
Catalog No. 50-005-82202 Supplier Med Vet International Supplier No. JOR710M
20mm Small Cruciate Needle. Swaged onto 80 lb. nylon leader line with 12mm crimp tube. 800mm L