Learn More
Med Vet International Cruciate Repair Kit, 2.7mm with 16mm x 2.0mm Coritcal Self-Tapping Screws, J1072FT
Catalog No. 5000583477
Encompass_Preferred
2.7mm screws hex head 2.0mm pilot2.7 self-tapping Cortical 16mm
Catalog No. 50-005-83477 Supplier Med Vet International Supplier No. JOR1072FT
2.7mm screws hex head 2.0mm pilot2.7 self-tapping Cortical 16mm