Learn More
Perkin Elmer US LLC Ceramic Sample Pans - Qty. 10
Catalog No. 509052221
Encompass_Preferred
These ceramic sample pans are designed to hold a variety of substances, both volatile and non-volatile, and to withstand temperatures ranging from sub-ambient to over 1000 °C.
Catalog No. 50-905-2221 Supplier Perkin Elmer US LLC Supplier No. N5370464
These ceramic sample pans are designed to hold a variety of substances, both volatile and non-volatile, and to withstand temperatures ranging from sub-ambient to over 1000 °C.