Learn More
Hach Company Crucible, Gooch, 40mL Capacity
Supplier:  Hach Company 2098641
Encompass
Perforated bottom, glazed inside and out except for outside bottom surface. Filter for Gooch Crucible available and sold separately.25-mL Gooch Crucible is recommended for use in Standard Methods 2540 C and 2540 D to determine Total Suspended Solids.
Catalog No. 50-199-5377