Learn More
Wilmad Labglass CRUCIBLE GOOCH LOW 30ML M
Supplier:  Wilmad Labglass LG5480108
Encompass_Preferred
Crucible Gooch Low; Unit has sealed-in fritted glass disc. For analytical work involving precipitation, drying, weighing. Avoid sudden temperature changes when heating or cooling. Fine, Medium, Coarse porosities available.
Catalog No. 50-806-175