Learn More
Velp™ Crucible Holder HU6 for SER 148 Solvent Extractor

Description
- For use with SER 148 Series Solvent Extractors
Specifications
Specifications
| Product Type | Crucible Holder |
| For Use With (Equipment) | SER 148 Series Solvent Extractor |
| Color | White |
| Voltage | 115/230 VAC |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.