Learn More
Cole-Parmer Essentials Tall-Form Crucible with Cover, porcelain, 10 mL, 6/pk
Catalog No. NC1860939 Shop All Cole-Parmer Products
Encompass_Preferred
Glazed on inner and outer surfaces, except outer bottom and rim. Withstand temperatures to 2102°F (1150°C) and heating rates up to 200°C per hour. Cover is included. Autoclavable.
Catalog No. NC1860939 Supplier Cole-Parmer Supplier No. 6310025
Glazed on inner and outer surfaces, except outer bottom and rim. Withstand temperatures to 2102°F (1150°C) and heating rates up to 200°C per hour. Cover is included. Autoclavable.