Learn More
Chemglass Life Sciences 25 ML capacity, top dia. 47 mm, base: 18 mm, height: 30mm
SDP
Catalog No. 50872002
Encompass_Preferred
Crucibles with Lids, Quartz, 25 ML capacity, top dia. 47 mm, base: 18 mm, height: 30mm. Crucible for use in temperatures up to 1000¿C. Perfect for use in muffle furnace ashing procedures. Quartz material ensures sample purity for further analysis.
Catalog No. 50-872-002 Supplier Chemglass Life Sciences Supplier No. CGQ400002
Crucibles with Lids, Quartz, 25 ML capacity, top dia. 47 mm, base: 18 mm, height: 30mm. Crucible for use in temperatures up to 1000¿C. Perfect for use in muffle furnace ashing procedures. Quartz material ensures sample purity for further analysis.