Learn More
Chemglass Life Sciences 15 ML capacity, top dia. 41mm, base: 15mm, height: 28mm
SDP
Catalog No. 501532076
Encompass_Preferred
15 ML capacity, top dia. 41mm, base: 15mm, height: 28mm. Crucible for use in temperatures up to 1000C. Perfect for use in muffle furnace ashing procedures. Quartz material ensures sample purity for further analysis.
Catalog No. 50-153-2076 Supplier Chemglass Life Sciences Supplier No. CGQ400001
15 ML capacity, top dia. 41mm, base: 15mm, height: 28mm. Crucible for use in temperatures up to 1000C. Perfect for use in muffle furnace ashing procedures. Quartz material ensures sample purity for further analysis.