Learn More
Chemglass Life Sciences 40 ML capacity, top dia. 57 mm, base: 20 mm, height: 45mm
SDP
Catalog No. 50872003
Encompass_Preferred
Crucibles with Lids, Quartz, 40 ML capacity, top dia. 57 mm, base: 20 mm, height: 45mm. Crucible for use in temperatures up to 1000¿C. Perfect for use in muffle furnace ashing procedures. Quartz material ensures sample purity for further analysis.
Catalog No. 50-872-003 Supplier Chemglass Life Sciences Supplier No. CGQ400003
Crucibles with Lids, Quartz, 40 ML capacity, top dia. 57 mm, base: 20 mm, height: 45mm. Crucible for use in temperatures up to 1000¿C. Perfect for use in muffle furnace ashing procedures. Quartz material ensures sample purity for further analysis.