Learn More
Chemglass Life Sciences 100 ML capacity, top dia. 62 mm, base: 35 mm, height: 67mm
SDP
Catalog No. 50872004
Encompass_Preferred
Crucibles with Lids, Quartz, 100 ML capacity, top dia. 62 mm, base: 35 mm, height: 67mm. Crucible for use in temperatures up to 1000¿C. Perfect for use in muffle furnace ashing procedures. Quartz material ensures sample purity for further analysis.
Catalog No. 50-872-004 Supplier Chemglass Life Sciences Supplier No. CGQ400004
Crucibles with Lids, Quartz, 100 ML capacity, top dia. 62 mm, base: 35 mm, height: 67mm. Crucible for use in temperatures up to 1000¿C. Perfect for use in muffle furnace ashing procedures. Quartz material ensures sample purity for further analysis.