Learn More
Electron Microscopy Sciences Crucible Tong, Bowed SS, 200 mm
SDP
Catalog No. 5032939
Encompass_Preferred
These tweezers are made to be used with precious metal crucibles. These tweezers are for use with precious metal crucibles. Platinum tips.
Catalog No. 50-329-39 Supplier Electron Microscopy Sciences Supplier No. 66510
These tweezers are made to be used with precious metal crucibles. These tweezers are for use with precious metal crucibles. Platinum tips.