Learn More
Electron Microscopy Sciences Crucible Tweezers, 130 mm
SDP
Catalog No. 5032938
Encompass_Preferred
These tweezers are made to be used with precious metal crucibles. These tongs are for use with precious metal crucibles. Platinum tips.
Catalog No. 50-329-38 Supplier Electron Microscopy Sciences Supplier No. 66511
These tweezers are made to be used with precious metal crucibles. These tongs are for use with precious metal crucibles. Platinum tips.