Learn More
LECO Corporation Crucible Cover - with 10 mm hole, for use with 528-018 and 528-038 Crucibles | 100/pack
Catalog No. NC9867636
Encompass
Crucible cover for CS WC CS / 230, 600 Series, 744 Series, 844 Series.
Catalog No. NC9867636 Supplier LECO Corporation Supplier No. 619880100
Crucible cover for CS WC CS / 230, 600 Series, 744 Series, 844 Series.