Learn More
Medchemexpress LLC HY-N6979 10mg Medchemexpress, Crustecdysone CAS:5289-74-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN697910MG
Encompass_Preferred
Medchemexpress, HY-N6979 10mg Crustecdysone CAS:5289-74-7 Crustecdysone (20-Hydroxyecdysone) is a naturally occurring ecdysteroid hormone isolated from Cyanotis arachnoides C.B.Clarke which controls the ecdysis (moulting) and metamorphosis of arthropods, it inhibits caspase activity and induces autophagy via the 20E nuclear receptor complex, EcR-USP.Crustecdysone exhibits regulatory or protective roles in the cardiovascular system. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0487