Learn More
Cayman Chemical CruzaIn-IN-1 10mg
SDP
Catalog No. 502917918
Encompass_Preferred
An inhibitor of cruzain (IC50 = 0.2 nM); selective for cruzain over cathepsin L-like protein and cathepsin B-like protease (IC50s = T. brucei (MIC50 = 25 µM)
Catalog No. 50-291-7918 Supplier Cayman Chemical Supplier No. 3735010 MG
An inhibitor of cruzain (IC50 = 0.2 nM); selective for cruzain over cathepsin L-like protein and cathepsin B-like protease (IC50s = T. brucei (MIC50 = 25 µM)