Learn More
Medchemexpress LLC HY-10836 10mg Medchemexpress, Cruzain-IN-1 CAS:1199523-24-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1083610MG
Encompass_Preferred
Medchemexpress, HY-10836 10mg Cruzain-IN-1 CAS:1199523-24-4 Cruzain-IN-1 is a covalent and reversible Cruzain inhibitor, with an IC50 of 10 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8539