Learn More
Mitegen Replacement Cryo Canister For IC Biomedical CX100, 11-Inch Length, 2.64" Diameter, Stainless Steel
SDP
Catalog No. NC3343926
Encompass
11-Inch Replacement Cryo Canister is for the IC Biomedical CX100 Cryo Express dry shipper, ensuring continued optimal performance for your cryogenic sample storage and transport. Constructed from durable, thin-gauge stainless steel, this canister provides a reliable and robust solution for securely holding various materials at cryogenic temperatures. Its precise dimensions, measuring 2.64 inches in diameter by 11 inches in length (67 x 279mm), ensure perfect compatibility and efficient use of space within your dry shipper. As an essential replacement component, it helps maintain the integrity and longevity of your CX100 dry shipper system, supporting extended holding times and providing a secure environment for your valuable samples. This canister is indispensable for laboratories relying on the CX100 Cryo Express dry shipper for safe and dependable cryogenic sample management.
Catalog No. NC3343926 Supplier Mitegen Supplier No. TWR0059C24
11-Inch Replacement Cryo Canister is for the IC Biomedical CX100 Cryo Express dry shipper, ensuring continued optimal performance for your cryogenic sample storage and transport. Constructed from durable, thin-gauge stainless steel, this canister provides a reliable and robust solution for securely holding various materials at cryogenic temperatures. Its precise dimensions, measuring 2.64 inches in diameter by 11 inches in length (67 x 279mm), ensure perfect compatibility and efficient use of space within your dry shipper. As an essential replacement component, it helps maintain the integrity and longevity of your CX100 dry shipper system, supporting extended holding times and providing a secure environment for your valuable samples. This canister is indispensable for laboratories relying on the CX100 Cryo Express dry shipper for safe and dependable cryogenic sample management.