Learn More
Aviva Systems Biology Corp CRY1 Antibody - N-terminal region (ARP59758_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59758_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRY1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CRY1. Alternate names: PHLL1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-815