Learn More
Aviva Systems Biology Corp CRYAB Antibody - C-terminal region (ARP48196_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48196_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CRYAB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CRYAB. Alternate names: CRYA2, CTPP2, HSPB5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-59751