Learn More
Aviva Systems Biology Corp Cryab Antibody - N-terminal region (ARP48195_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48195_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cryab. It was validated on Western Blot. . Alternate names: AACRYAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-285-352