Learn More
Enzo Life Sciences alphaB Crystallin (human), (recombinant) (His-tag) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPP228050
Encompass_Preferred
Alternative name: CRYAB, HspB5, Heat shock protein beta 5. Purity: ≥95% (SDS-PAGE; Western blot). MW: ~26kDa (observed). Formulation: Liquid. In 50mM TRIS, pH 7.5, containing 50mM sodium chloride and 1mM EDTA. Source: Produced in E. coli. Human αB-Crystallin is fused at the N-terminus to a His-tag. Long Term Storage: -80°C. Applications: WB .
Catalog No. 50-200-7598