Learn More
Enzo Life Sciences alphaB Crystallin monoclonal antibody (1B6.1 3G4) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS7060200
Encompass_Preferred
Clone: 1B6.1-3G4. Isotype: IgG1. Immunogen: Bovine αB-crystallin. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat Chicken, Bovine. Applications: ICC, IF, IHC (PS), WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-3245