Learn More
Enzo Life Sciences alphaB Crystallin (pSer19) polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADISPA225E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human αB-crystallin. The sequence is completely conserved in mouse, rat and bovine phosphorylated at Ser19. UniProt ID: P02511. Source: Purified from rabbit serum. Species reactivity: Human, Mouse Bovine. Applications: IP, WB. Recommended Dilutions: Immunoprecipitation (12.5µg/ml) Western Blot (1µg/ml, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. In sodium phosphate buffer, pH 7.0, containing 3% BSA.
Catalog No. 50-200-7422