Learn More
Enzo Life Sciences alphaB Crystallin (pSer45) polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA226F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human αB-crystallin phosphorylated at Ser45. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Xenopus, Scallop, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: IP, WB. Purity: Affinity purified. Formulation: Liquid. In 0.1M sodium phosphate buffer, pH 7.0, containing 1.5% BSA and 0.1% sodium azide.
Catalog No. 50-200-7424