Learn More
Enzo Life Sciences alphaB Crystallin (pSer45) polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA226D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human αB-crystallin phosphorylated at Ser45. UniProt ID: P02511. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Xenopus, Scallop, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: IP, WB. Recommended Dilutions: Western Blot (1:500, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Affinity purified. Formulation: Liquid. In 0.1M sodium phosphate buffer, pH 7.0, containing 1.5% BSA and 0.1% sodium azide.
Catalog No. 50-200-7423