Learn More
Enzo Life Sciences alphaB Crystallin (pSer59) polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA227F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to to aa 54-64 of of human αB-crystallin phosphorylated at Ser59. Source: Purified from rabbit serum. Species reactivity: Mouse, Rat, Sheep, Porcine, Bovine. Applications: IP, WB. Formulation: Liquid. In sodium phosphate buffer, pH 7.0, containing 0.1% sodium azide, 3% BSA, and 50% glycerol.
Catalog No. 50-200-7425