Learn More
Cole-Parmer Argos Technologies PolarSafe Aluminum CyroCane, with Platforms, 5 Vials; 12/PK
Catalog No. 501960881 Shop All Cole-Parmer Products
Encompass_Preferred
These aluminum storage canes are designed to store cryogenic vials in Dewars or liquid nitrogen tanks. These canes will accommodate most cryogenic vials in 1.0-, 1.2-, 1.5-, 1.8-, and 2.0-mL sizes. The cane without vial platform can also accommodate 5.0 mL vials. Choose from two styles, without or without vial platforms. Each can accommodate 5 vials. The clear plastic sleeve keeps your vials in place while being stored on the cane.
Catalog No. 50-196-0881 Supplier Cole-Parmer Supplier No. 2441003
These aluminum storage canes are designed to store cryogenic vials in Dewars or liquid nitrogen tanks. These canes will accommodate most cryogenic vials in 1.0-, 1.2-, 1.5-, 1.8-, and 2.0-mL sizes. The cane without vial platform can also accommodate 5.0 mL vials. Choose from two styles, without or without vial platforms. Each can accommodate 5 vials. The clear plastic sleeve keeps your vials in place while being stored on the cane.