Learn More
Description
- Two variations available:, Cryer Elevator 44 Left, Cryer Elevator 45 Right
- Sharp triangular blade ideal for elevating broken roots and difficult posterior teeth
- Designed for rotational leverage to efficiently remove root fragments
- Strong, rigid construction withstands heavy lifting forces
- Ergonomic handle ensures secure, slip-resistant grip
- Precision-machined from high-grade surgical stainless steel
- Corrosion-resistant and fully autoclavable for repeated use
- Excellent access for maxillary and mandibular root removal
- Applications: Dental extractions, oral surgery, forensic dentistry, and autopsy/postmortem root elevation, mandible access, and tooth removal
Specifications
Specifications
| Product Type | Cryer Elevator |
| Handle Material | Stainless Steel |
| Length (Metric) | 15.5 cm |
| Material | Stainless Steel |
| Size | Adult |
| Autoclavable | Autoclavable |
| Color | Silver |
| Tip Style | Left |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.