Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Cryer Elevators

Catalog No. 21124707 Shop All Surgical Design Products
Change view
Click to view available options
Tip Style:
Left
Right
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
21-124-707 Right
21-124-706 Left
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-124-707 Supplier Surgical Design™ Supplier No. DELCRY45R
Add to Cart
Add to Cart

Cryer elevators provide powerful leverage for removing fractured roots and impacted teeth in dental and surgical procedures.

  • Two variations available:, Cryer Elevator 44 Left, Cryer Elevator 45 Right
  • Sharp triangular blade ideal for elevating broken roots and difficult posterior teeth
  • Designed for rotational leverage to efficiently remove root fragments
  • Strong, rigid construction withstands heavy lifting forces
  • Ergonomic handle ensures secure, slip-resistant grip
  • Precision-machined from high-grade surgical stainless steel
  • Corrosion-resistant and fully autoclavable for repeated use
  • Excellent access for maxillary and mandibular root removal
  • Applications: Dental extractions, oral surgery, forensic dentistry, and autopsy/postmortem root elevation, mandible access, and tooth removal

Specifications

Product Type Cryer Elevator
Handle Material Stainless Steel
Length (Metric) 15.5 cm
Material Stainless Steel
Size Adult
Autoclavable Autoclavable
Color Silver
Tip Style Right
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.