Learn More
Enzo Life Sciences gamma Crystallin (bovine), (native) (1mg)
SDP
Supplier:  Enzo Life Sciences ADISPP240J
Encompass_Preferred
Alternative name: CRYGD. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~19kDa. Formulation: Liquid. In PBS containing 0.09% sodium azide. Source: Isolated from bovine eye lens. Long Term Storage: -80°C. Applications: WB .
Catalog No. 50-200-7602