Learn More
GA International Dual Point Cryogenic Waterproof Permanent Marker Red 6/pack
Catalog No. NC2394834
Encompass_Preferred
Waterproof, cryogenic permanent marker with dual point, fine and extra-fine high-quality tips. This 2-in-1 marker is available in a range of colors, and has excellent performance on plastic, glass, metal, and cloth surfaces. Ideal for use in bio-medical laboratories, this permanent marker can be used to write on cryogenic labels, for subsequent storage in liquid nitrogen, and lab freezers.
Catalog No. NC2394834 Supplier GA International Supplier No. MP1RE6
Waterproof, cryogenic permanent marker with dual point, fine and extra-fine high-quality tips. This 2-in-1 marker is available in a range of colors, and has excellent performance on plastic, glass, metal, and cloth surfaces. Ideal for use in bio-medical laboratories, this permanent marker can be used to write on cryogenic labels, for subsequent storage in liquid nitrogen, and lab freezers.